Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 25g, 3-Fluoro-5-methoxyphenylboronic acid, 98%, [609807-25-2], F2100 169.95 C7H8BFO3

Catalog No. 5037048
Encompass_Preferred

Compound: 3-Fluoro-5-methoxyphenylboronic acid

CAS#: 609807-25-2

MDL#: not listed

Background: A fluorinated, methoxylated phenylboronic acid used as a Suzuki-Miyaura coupling partner in pharmaceutical research. It is a common building block for constructing substituted biaryl scaffolds in medicinal chemistry.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-370-48 Supplier Frontier Specialty Chemicals Supplier No. F210025G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 3-Fluoro-5-methoxyphenylboronic acid

CAS#: 609807-25-2

MDL#: not listed

Background: A fluorinated, methoxylated phenylboronic acid used as a Suzuki-Miyaura coupling partner in pharmaceutical research. It is a common building block for constructing substituted biaryl scaffolds in medicinal chemistry.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.