Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 25g, 4-Bromo-3-formylindole, >98%, [98600-34-1], B1568 224.06 C9H6BrNO

Catalog No. 50981274
Encompass_Preferred

Compound: 4-Bromo-3-formylindole

CAS#: 98600-34-1

MDL#: MFCD05864695

Background: A brominated indole carbaldehyde used as an intermediate in medicinal chemistry synthesis. The bromine substituent provides a reactive site for cross-coupling, while the aldehyde allows further derivatization in the construction of substituted indole scaffolds.

Storage: Store at 2-8°C, under dry conditions.

Catalog No. 50-981-274 Supplier Frontier Specialty Chemicals Supplier No. B156825G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 4-Bromo-3-formylindole

CAS#: 98600-34-1

MDL#: MFCD05864695

Background: A brominated indole carbaldehyde used as an intermediate in medicinal chemistry synthesis. The bromine substituent provides a reactive site for cross-coupling, while the aldehyde allows further derivatization in the construction of substituted indole scaffolds.

Storage: Store at 2-8°C, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.