Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 25g, 4-Methoxypyridine-3-boronic acid hydrate, >97%, [1256355-26-6], M1192 1795%9 C6H10BNO4

Catalog No. 50982671
Encompass_Preferred

Compound: 4-Methoxypyridine-3-boronic acid hydrate

CAS#: 1256355-26-6

MDL#: MFCD07186213

Background: A methoxy-substituted pyridine boronic acid used as a Suzuki-Miyaura coupling partner in heterocyclic synthesis. It has also found use in deuteration reaction studies, serving as a substrate for investigating isotope-labeling methodology.

Storage: Store at 2-8°C, under dry conditions.

Catalog No. 50-982-671 Supplier Frontier Specialty Chemicals Supplier No. M119225G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 4-Methoxypyridine-3-boronic acid hydrate

CAS#: 1256355-26-6

MDL#: MFCD07186213

Background: A methoxy-substituted pyridine boronic acid used as a Suzuki-Miyaura coupling partner in heterocyclic synthesis. It has also found use in deuteration reaction studies, serving as a substrate for investigating isotope-labeling methodology.

Storage: Store at 2-8°C, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.