Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 25g, 4-(tert-Butoxycarbonyl)phenylboronic acid pinacol ester, 98%, [850568-72-8], B4499 304.2 C17H25BO4

Catalog No. 50981655
Encompass_Preferred

Compound: 4-(tert-Butoxycarbonyl)phenylboronic acid pinacol ester

CAS#: 850568-72-8

MDL#: MFCD03453055

Background: A Boc-protected ester-substituted phenylboronic ester used as a proteomics research reagent and Suzuki-Miyaura coupling partner. The protected carboxylate group allows for later deprotection and further functionalization in medicinal chemistry synthesis.

Storage: Store at 0-8°C, under dry conditions.

Catalog No. 50-981-655 Supplier Frontier Specialty Chemicals Supplier No. B449925G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 4-(tert-Butoxycarbonyl)phenylboronic acid pinacol ester

CAS#: 850568-72-8

MDL#: MFCD03453055

Background: A Boc-protected ester-substituted phenylboronic ester used as a proteomics research reagent and Suzuki-Miyaura coupling partner. The protected carboxylate group allows for later deprotection and further functionalization in medicinal chemistry synthesis.

Storage: Store at 0-8°C, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.