Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 25g, 5-Bromo-2-chloropyrimidine, 98%, [32779-36-5], B1216 193.43 C4H2BrClN2

Catalog No. 50981206
Encompass_Preferred

Compound: 5-Bromo-2-chloropyrimidine

CAS#: 32779-36-5

MDL#: MFCD00483232

Background: 5-Bromo-2-chloropyrimidine is used as a pharmaceutical intermediate in the preparation of macitentan, an endothelin receptor antagonist used to treat pulmonary hypertension. It also serves as a key intermediate in the synthesis of antiviral and anticancer agents, and is used in the formulation of herbicides and pesticides.

Catalog No. 50-981-206 Supplier Frontier Specialty Chemicals Supplier No. B121625G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 5-Bromo-2-chloropyrimidine

CAS#: 32779-36-5

MDL#: MFCD00483232

Background: 5-Bromo-2-chloropyrimidine is used as a pharmaceutical intermediate in the preparation of macitentan, an endothelin receptor antagonist used to treat pulmonary hypertension. It also serves as a key intermediate in the synthesis of antiviral and anticancer agents, and is used in the formulation of herbicides and pesticides.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.