Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 25g, 5-Bromo-2-cyanopyridine, 97%, [97483-77-7], B1626 183.01 C6H3BrN2

Catalog No. 50981313
Encompass_Preferred

Compound: 5-Bromo-2-cyanopyridine

CAS#: 97483-77-7

MDL#: MFCD00234144

Background: 5-Bromo-2-cyanopyridine serves as a key intermediate in the synthesis of anti-cancer and anti-inflammatory pharmaceutical candidates, including aza-terphenyl diamidine analogs with antiprotozoal activity. It is widely used in Suzuki-Miyaura coupling reactions and in the synthesis of pyridine-diketopyrrolopyrrole building blocks for low-band-gap copolymers used as electron donors in polymer solar cells.

Catalog No. 50-981-313 Supplier Frontier Specialty Chemicals Supplier No. B162625G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 5-Bromo-2-cyanopyridine

CAS#: 97483-77-7

MDL#: MFCD00234144

Background: 5-Bromo-2-cyanopyridine serves as a key intermediate in the synthesis of anti-cancer and anti-inflammatory pharmaceutical candidates, including aza-terphenyl diamidine analogs with antiprotozoal activity. It is widely used in Suzuki-Miyaura coupling reactions and in the synthesis of pyridine-diketopyrrolopyrrole building blocks for low-band-gap copolymers used as electron donors in polymer solar cells.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.