Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 25g, 5-Formyl-2-methoxypyridine, >98%, [65873-72-5], M1679 137.14 C7H7NO2

Catalog No. 50982694
Encompass_Preferred

Compound: 5-Formyl-2-methoxypyridine

CAS#: 65873-72-5

MDL#: MFCD02683446

Background: 5-Formyl-2-methoxypyridine is used as a substrate in the synthesis of flavanones via L-proline-catalyzed condensation with o-hydroxyarylketones. It also serves as an intermediate for the synthesis of somatostatin sst3 receptor ligands and as a building block for heterocyclic compounds in pharmaceutical research.

Catalog No. 50-982-694 Supplier Frontier Specialty Chemicals Supplier No. M167925G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 5-Formyl-2-methoxypyridine

CAS#: 65873-72-5

MDL#: MFCD02683446

Background: 5-Formyl-2-methoxypyridine is used as a substrate in the synthesis of flavanones via L-proline-catalyzed condensation with o-hydroxyarylketones. It also serves as an intermediate for the synthesis of somatostatin sst3 receptor ligands and as a building block for heterocyclic compounds in pharmaceutical research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.