Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 25g, 5-Formylpyrimidine, >97%, [10070-92-5], P1680 108.1 C5H4N2O

Catalog No. 5039000
Encompass_Preferred

Compound: 5-Formylpyrimidine

CAS#: 10070-92-5

MDL#: not listed

Background: A pyrimidine carbaldehyde used as a building block in pharmaceutical research. The aldehyde group provides a reactive handle for reductive amination and other condensation reactions in heterocyclic synthesis.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-390-00 Supplier Frontier Specialty Chemicals Supplier No. P168025G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 5-Formylpyrimidine

CAS#: 10070-92-5

MDL#: not listed

Background: A pyrimidine carbaldehyde used as a building block in pharmaceutical research. The aldehyde group provides a reactive handle for reductive amination and other condensation reactions in heterocyclic synthesis.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.