Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 25g, 6-Bromo-1H-indazole, 98%, [79762-54-2], B11052 197.032 C7H5BrN2

Catalog No. 5035455
Encompass_Preferred

Compound: 6-Bromo-1H-indazole

CAS#: 79762-54-2

MDL#: not listed

Background: A brominated indazole used as a building block in pharmaceutical research. Indazoles are privileged nitrogen heterocyclic scaffolds in drug discovery, and the bromine substituent provides a reactive site for cross-coupling functionalization.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-354-55 Supplier Frontier Specialty Chemicals Supplier No. B1105225G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 6-Bromo-1H-indazole

CAS#: 79762-54-2

MDL#: not listed

Background: A brominated indazole used as a building block in pharmaceutical research. Indazoles are privileged nitrogen heterocyclic scaffolds in drug discovery, and the bromine substituent provides a reactive site for cross-coupling functionalization.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.