Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 25g, Allylboronic acid pinacol ester, 97%, [72824-04-5], A1648 168.05 C9H17BO2

Catalog No. 50981036
Encompass_Preferred

Compound: Allylboronic acid pinacol ester

CAS#: 72824-04-5

MDL#: MFCD00013347

Background: Allylboronic acid pinacol ester is a synthetic specialty chemical useful in the synthesis of pharmaceuticals and specialty organic chemicals. It is commonly used as an allylating reagent in organic synthesis and in transition metal-catalyzed coupling reactions.

Storage: Store at 2-8°C, under dry conditions.

Catalog No. 50-981-036 Supplier Frontier Specialty Chemicals Supplier No. A164825G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: Allylboronic acid pinacol ester

CAS#: 72824-04-5

MDL#: MFCD00013347

Background: Allylboronic acid pinacol ester is a synthetic specialty chemical useful in the synthesis of pharmaceuticals and specialty organic chemicals. It is commonly used as an allylating reagent in organic synthesis and in transition metal-catalyzed coupling reactions.

Storage: Store at 2-8°C, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.