Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 25g, Bis(neopentyl glycolato)diboron, 98%, [201733-56-4], B5100 225.89 C10H20B2O4

Catalog No. 50981666
Encompass_Preferred

Compound: Bis(neopentyl glycolato)diboron

CAS#: 201733-56-4

MDL#: MFCD02093062

Background: Bis(neopentyl glycolato)diboron is a diboron reagent used to prepare boronic acid esters via Miyaura borylation. It enables Suzuki-Miyaura coupling reactions and other palladium-catalyzed transformations with precise control over borylation. It has also been explored as an intermediate for boron-based tumor-targeting compounds.

Catalog No. 50-981-666 Supplier Frontier Specialty Chemicals Supplier No. B510025G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: Bis(neopentyl glycolato)diboron

CAS#: 201733-56-4

MDL#: MFCD02093062

Background: Bis(neopentyl glycolato)diboron is a diboron reagent used to prepare boronic acid esters via Miyaura borylation. It enables Suzuki-Miyaura coupling reactions and other palladium-catalyzed transformations with precise control over borylation. It has also been explored as an intermediate for boron-based tumor-targeting compounds.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.