Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 25g, Dibenzofuran-4-boronic acid, 97%, [100124-06-9], D5438 212.01 C12H9BO3

Catalog No. 5036743
Encompass_Preferred

Compound: Dibenzofuran-4-boronic acid

CAS#: 100124-06-9

MDL#: not listed

Background: A dibenzofuran-substituted boronic acid used as a Suzuki-Miyaura coupling partner in materials science and pharmaceutical research. Dibenzofuran-containing building blocks are commonly used in the synthesis of organic electronic and photoluminescent materials.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-367-43 Supplier Frontier Specialty Chemicals Supplier No. D543825G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: Dibenzofuran-4-boronic acid

CAS#: 100124-06-9

MDL#: not listed

Background: A dibenzofuran-substituted boronic acid used as a Suzuki-Miyaura coupling partner in materials science and pharmaceutical research. Dibenzofuran-containing building blocks are commonly used in the synthesis of organic electronic and photoluminescent materials.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.