Learn More
Biotage Biotage Sfar Duo column, 25G Frit 20/PK
SDP
Catalog No. NC1777273
Encompass
Frit used to capture dried samples inside 25 gram and 30 gram Biotage Sfar Duo columns. These are used when the Sfar Duo cap is removed and sample is placed on top of the column bed. The frits are placed and the cap is attached and sealed to the top of the column.
Catalog No. NC1777273 Supplier Biotage Supplier No. 415262SP
Frit used to capture dried samples inside 25 gram and 30 gram Biotage Sfar Duo columns. These are used when the Sfar Duo cap is removed and sample is placed on top of the column bed. The frits are placed and the cap is attached and sealed to the top of the column.