Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 25g, Methylboronic acid, 98%, [13061-96-6], M7469 59.86 CH5BO2

Catalog No. 50982804
Encompass_Preferred

Compound: Methylboronic acid

CAS#: 13061-96-6

MDL#: MFCD00002105

Background: Methylboronic acid is a simple alkylboronic acid used in Suzuki and Stille cross-coupling reactions to form carbon-carbon bonds in palladium-catalyzed synthesis. It is used in the development of boron-containing pharmaceuticals and agrochemicals, including chrysin analogs and casein kinase I inhibitors. It also serves as a reagent for derivatizing carbohydrates and other biologically active compounds.

Catalog No. 50-982-804 Supplier Frontier Specialty Chemicals Supplier No. M746925G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: Methylboronic acid

CAS#: 13061-96-6

MDL#: MFCD00002105

Background: Methylboronic acid is a simple alkylboronic acid used in Suzuki and Stille cross-coupling reactions to form carbon-carbon bonds in palladium-catalyzed synthesis. It is used in the development of boron-containing pharmaceuticals and agrochemicals, including chrysin analogs and casein kinase I inhibitors. It also serves as a reagent for derivatizing carbohydrates and other biologically active compounds.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.