Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 25g, Pyridine-3-boronic acid, 95%, [1692-25-7], P5775 122.92 C5H6BNO2

Catalog No. 50982974
Encompass_Preferred

Compound: Pyridine-3-boronic acid

CAS#: 1692-25-7

MDL#: MFCD00674177

Background: Pyridine-3-boronic acid is a heteroaryl boronic acid used as a building block in Suzuki-Miyaura cross-coupling reactions to construct biaryl compounds containing a pyridine ring. It has been applied in phosphine-free and regioselective coupling methods, including tandem coupling-annulation sequences to build fused nitrogen heterocycles. It also supplies the 3-pyridyl unit used in the synthesis of the Bcr-Abl tyrosine-kinase inhibitor nilotinib.

Catalog No. 50-982-974 Supplier Frontier Specialty Chemicals Supplier No. P577525G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: Pyridine-3-boronic acid

CAS#: 1692-25-7

MDL#: MFCD00674177

Background: Pyridine-3-boronic acid is a heteroaryl boronic acid used as a building block in Suzuki-Miyaura cross-coupling reactions to construct biaryl compounds containing a pyridine ring. It has been applied in phosphine-free and regioselective coupling methods, including tandem coupling-annulation sequences to build fused nitrogen heterocycles. It also supplies the 3-pyridyl unit used in the synthesis of the Bcr-Abl tyrosine-kinase inhibitor nilotinib.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.