Learn More
Cayman Chemical 25-hydroxy Vitamin D3 3,3’-aminopropyl ether, Purity: ≥98%, CAS: 163018-26-6, 10 mg
SDP
Catalog No. 502995565
Encompass_Preferred
A vitamin D3 analog; contains a highly reactive amine group that can be coupled to a sepharose matrix for affinity purification of vitamin D binding protein; inhibits 25-hydroxy vitamin D3-1α-hydroxylase
Catalog No. 50-299-5565 Supplier Cayman Chemical Supplier No. 2312210 MG
A vitamin D3 analog; contains a highly reactive amine group that can be coupled to a sepharose matrix for affinity purification of vitamin D binding protein; inhibits 25-hydroxy vitamin D3-1α-hydroxylase