Learn More
Cayman Chemical 25hydroxy VItamIn D3 3 3am 1mg
SDP
Catalog No. 502995564
Encompass_Preferred
A vitamin D3 analog; contains a highly reactive amine group that can be coupled to a sepharose matrix for affinity purification of vitamin D binding protein; inhibits 25-hydroxy vitamin D3-1α-hydroxylase
Catalog No. 50-299-5564 Supplier Cayman Chemical Supplier No. 231221 MG
Add to Cart
Add to Cart
A vitamin D3 analog; contains a highly reactive amine group that can be coupled to a sepharose matrix for affinity purification of vitamin D binding protein; inhibits 25-hydroxy vitamin D3-1α-hydroxylase