Learn More
Cayman Chemical 25hydroxy VItamIn D3 3 3am 5mg
SDP
Catalog No. 502994891
Encompass_Preferred
A vitamin D3 analog; contains a highly reactive amine group that can be coupled to a sepharose matrix for affinity purification of vitamin D binding protein; inhibits 25-hydroxy vitamin D3-1α-hydroxylase
Catalog No. 50-299-4891 Supplier Cayman Chemical Supplier No. 231225 MG
A vitamin D3 analog; contains a highly reactive amine group that can be coupled to a sepharose matrix for affinity purification of vitamin D binding protein; inhibits 25-hydroxy vitamin D3-1α-hydroxylase