Learn More
Cayman Chemical 25I-NBMD (hydrochloride), Purity: ≥98%, CAS: 1539266-14-2, 10 mg
SDP
Catalog No. 502953882
Encompass_Preferred
A derivative of the hallucinogen 2C-I with high affinity (Ki = 0.049–0.21 nM) and activity (EC50 = 8.2 nM) at the serotonin receptor 5-HT2A; intended for research and forensic applications
Catalog No. 50-295-3882 Supplier Cayman Chemical Supplier No. 1194710 MG
A derivative of the hallucinogen 2C-I with high affinity (Ki = 0.049–0.21 nM) and activity (EC50 = 8.2 nM) at the serotonin receptor 5-HT2A; intended for research and forensic applications