Learn More
Cayman Chemical 25I-NBOH (hydrochloride), Purity: ≥95%, CAS: 1539266-12-0, 5 mg
SDP
Catalog No. 502970754
Encompass_Preferred
A derivative of 2C-I that acts as a potent agonist for the human serotonin 5-HT2A receptor (Ki = 0.061 nM); intended for research and forensic applications
Catalog No. 50-297-0754 Supplier Cayman Chemical Supplier No. 149095 MG
A derivative of 2C-I that acts as a potent agonist for the human serotonin 5-HT2A receptor (Ki = 0.061 nM); intended for research and forensic applications