Learn More
ATLANTIC BIOPHARM SOLUTIONS LLC .25IN SB SERIES, BRAIDED USP
Catalog No. NC2229067
Encompass_Preferred
.25in SB Series, Braided USPClass VI Silicone Assy w/.5in 316L ElectropolishedReusable Mini Tri Clamp Each End 39 IN OAL. Qte 8498
This item is not returnable. View return policy
Catalog No. NC2229067 Supplier ATLANTIC BIOPHARM SOLUTIONS LLC Supplier No. 04RFSB08RTC08RTCSSEP 39
.25in SB Series, Braided USPClass VI Silicone Assy w/.5in 316L ElectropolishedReusable Mini Tri Clamp Each End 39 IN OAL. Qte 8498