Learn More
Cayman Chemical 25INBOMe 3methoxy Isomerh 10mg
SDP
Catalog No. 502971475
Encompass_Preferred
Differs from 25I-NBOMe by having a methoxy group at the three, rather than two, position on the benzene ring; intended for research and forensic applications
Catalog No. 50-297-1475 Supplier Cayman Chemical Supplier No. 1478410 MG
Differs from 25I-NBOMe by having a methoxy group at the three, rather than two, position on the benzene ring; intended for research and forensic applications