Learn More
Cayman Chemical 25I-NBOMe 4-methoxy isomer (hydrochloride), Purity: ≥98%, CAS: 1566571-64-9, 10 mg
SDP
Catalog No. 502971025
Encompass_Preferred
Differs from 25I-NBOMe by having a methoxy group at the four, rather than two, position on the benzene ring; intended for research and forensic applications
Catalog No. 50-297-1025 Supplier Cayman Chemical Supplier No. 1478510 MG
Differs from 25I-NBOMe by having a methoxy group at the four, rather than two, position on the benzene ring; intended for research and forensic applications