Learn More
Neta Scientific Agilent 250ul glass inserts, deactivated 100/PK
SDP
Catalog No. 50929401
Encompass_Preferred
This item is manufactured by Agilent
Agilent Vial insert, 250 uL, deactivated glass with polymer feet, 100/pk Insert size: 5.6 x 30 mm - High Recovery Vials & Inserts
UNSPSC Code  41121806
- 58
Catalog No. 50-929-401 Supplier Neta Scientific Supplier No. 51818872
This item is manufactured by Agilent
Agilent Vial insert, 250 uL, deactivated glass with polymer feet, 100/pk Insert size: 5.6 x 30 mm - High Recovery Vials & Inserts
UNSPSC Code  41121806
- 58