Learn More
AirSea USA 25L UN steel drum with lacquer lining and 51mm bung. UN Mark: 1A1/X/550. This 25L steel drum is lacquer lined and approved for shipping dangerous goods as liquids within Packing Groups I, II & III.
Catalog No. NC2101141
Encompass_Preferred
25L UN steel drum with lacquer lining and 51mm bung. UN Mark: 1A1/X/550. This 25L steel drum is lacquer lined and approved for shipping dangerous goods as liquids within Packing Groups I, II & III.
This item is not returnable. View return policy
Catalog No. NC2101141 Supplier AirSea USA Supplier No. 16
25L UN steel drum with lacquer lining and 51mm bung. UN Mark: 1A1/X/550. This 25L steel drum is lacquer lined and approved for shipping dangerous goods as liquids within Packing Groups I, II & III.