Learn More
AirSea USA 25L UN Steel Drum Lacquer Lined, 1A2/X53/S
Catalog No. 501534830
Encompass_Preferred
25L UN Steel Drum - Lacquer Lined. UN Mark: 1A2/X53/S. This 25L steel drum is approved to ship dangerous goods as solids in packing groups I, II, III.
Catalog No. 50-153-4830 Supplier AirSea USA Supplier No. 780
25L UN Steel Drum - Lacquer Lined. UN Mark: 1A2/X53/S. This 25L steel drum is approved to ship dangerous goods as solids in packing groups I, II, III.