Learn More
Avanti Polar Lipids 18:1 Lyso PC, 19420-56-5, MFCD00133435, powder, 25mg
Catalog No. NC1385980
Encompass
Molecular Formula C26H52NO7P, Purity >99% LPC; may contain up to 10% of the 2-LPC isomer, Formula Weight 521.667, Synonyms: 1-oleoyl-2-hydroxy-sn-glycero-3-phosphocholine
Catalog No. NC1385980 Supplier Avanti Polar Lipids Supplier No. 845875P25MG
Molecular Formula C26H52NO7P, Purity >99% LPC; may contain up to 10% of the 2-LPC isomer, Formula Weight 521.667, Synonyms: 1-oleoyl-2-hydroxy-sn-glycero-3-phosphocholine