Learn More
Cayman Chemical Vandetanib, 443913-73-3, 25 mg
SDP
Supplier:  Cayman Chemical 1470625
Encompass_Preferred
A broad spectrum, orally available kinase inhibitor that targets primarily tyrosine kinases, including VEGFR and EGFR, with IC50 values in the nanomolar range; also potently blocks non-receptor tyrosine kinases, including ABL, RET, and SRC, as well as several serine/threonine kinases
Catalog No. NC0706691
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.