Learn More
IBA LifeSciences 10x Buffer E - Strep-Tactin Elution Buffer with Desthiobiotin; 25 ml
Catalog No. NC9261075
Encompass_Preferred
10x Buffer E is a 10x concentrated Strep-Tactin elution buffer. The contained desthiobiotin is a modified form of biotin that displaces Strep-tagII and Twin-Strep-tag fusion proteins from the modified biotin-binding pocket of Strep-Tactin in a competitive manner, resulting in a mild elution. Please note that 10x Buffer E is not applicable for elution from Strep-TactinXT resins.
Catalog No. NC9261075 Supplier IBA LifeSciences Supplier No. 21000025
10x Buffer E is a 10x concentrated Strep-Tactin elution buffer. The contained desthiobiotin is a modified form of biotin that displaces Strep-tagII and Twin-Strep-tag fusion proteins from the modified biotin-binding pocket of Strep-Tactin in a competitive manner, resulting in a mild elution. Please note that 10x Buffer E is not applicable for elution from Strep-TactinXT resins.