Learn More
Ace Glass, Inc. 25mL microscale, pear-shaped flask with two 14/10 joints externally threaded for O-ring cap seal. Supplied with holed caps, PTFE septa, and FETFE O-rings.
SDP
Catalog No. 50976929
Encompass_Preferred
Catalog No. 50-976-929 Supplier Ace Glass, Inc. Supplier No. 947929