Learn More
Ace Glass, Inc. 25mL graduated cylindrical funnel, 14/20 top, 14/20 bottom, PTFE stopcock plug
SDP
Catalog No. 50976958
Encompass_Preferred
25mL graduated cylindrical funnel with 14/20 top and bottom joints, a drip stem, and double scale graduations that are calibrated from the stopcock. Drip stem does not extend beyond the joint, thereby eliminating a possible source of breakage. Funnel has a PTFE stopcock plug.
Catalog No. 50-976-958 Supplier Ace Glass, Inc. Supplier No. 949905
25mL graduated cylindrical funnel with 14/20 top and bottom joints, a drip stem, and double scale graduations that are calibrated from the stopcock. Drip stem does not extend beyond the joint, thereby eliminating a possible source of breakage. Funnel has a PTFE stopcock plug.