Learn More
Chemglass Life Sciences 25mL 2-Neck Pear Shaped Flask, 1-CN 14/20 Outer, 1-SN 10/18 Outer Angled 20°
SDP
Catalog No. CG155809
Encompass_Preferred
25mL 2-Neck Pear Shaped Flask, 1-CN 14/20 Outer, 1-SN 10/18 Outer Angled 20¿. Pear shape, HEAVY WALL 2-neck flask with standard taper outer joints. Flasks with a 10/18 outer joint on the side neck are used with either jointed thermometers or CG-1079 inlet adapters.
Catalog No. CG155809 Supplier Chemglass Life Sciences Supplier No. CG155809
25mL 2-Neck Pear Shaped Flask, 1-CN 14/20 Outer, 1-SN 10/18 Outer Angled 20¿. Pear shape, HEAVY WALL 2-neck flask with standard taper outer joints. Flasks with a 10/18 outer joint on the side neck are used with either jointed thermometers or CG-1079 inlet adapters.