Learn More
Sterlitech Corporation Sterlitech Silver Membrane, 0.45 Micron, 25mm, 50/Pk
SDP
Catalog No. NC9039914
Encompass_Preferred
Pure metallic silver (99.97% pure silver), hydrophilic membrane filter, pore rating of 0.45 microns and a diameter of 25 mm
Catalog No. NC9039914 Supplier Sterlitech Corporation Supplier No. 45335
Add to Cart
Add to Cart
Pure metallic silver (99.97% pure silver), hydrophilic membrane filter, pore rating of 0.45 microns and a diameter of 25 mm