Learn More
Chemglass Life Sciences 25mL Addition Funnel, Graduated, 24/40 Joint Size, 2mm PTFE Metering Stpk, 240mm OAH
SDP
Catalog No. CG171306
Encompass_Preferred
25mL Addition Funnel, Graduated, 24/40 Joint Size, 2mm PTFE Metering Stopcock, 240mm OAH. Cylindrical style addition funnel with a PTFE adjustable bore metering plug. Turning the adjustment knob allows flow rate to be controlled from dropwise to full flow. For replacement stoppers see CG-3000.
Catalog No. CG171306 Supplier Chemglass Life Sciences Supplier No. CG171306
25mL Addition Funnel, Graduated, 24/40 Joint Size, 2mm PTFE Metering Stopcock, 240mm OAH. Cylindrical style addition funnel with a PTFE adjustable bore metering plug. Turning the adjustment knob allows flow rate to be controlled from dropwise to full flow. For replacement stoppers see CG-3000.