Learn More
GREENWOOD PRODUCTS 25ml Amber Glass Buret
SDP
Catalog No. NC3660224
Encompass_Preferred
Automatic Buret, 25mL, Amber, Glass Stopcock, Automatic Self-Zero, Class A, Graduated (w/ 1000mL Bottle, Rubber Bulb) (ea)
Catalog No. NC3660224 Supplier GREENWOOD PRODUCTS Supplier No. TG1830234
Automatic Buret, 25mL, Amber, Glass Stopcock, Automatic Self-Zero, Class A, Graduated (w/ 1000mL Bottle, Rubber Bulb) (ea)