Learn More
CELLTREAT Scientific Products 25mL Centrifuge Tube, Snap-Pop Lid - Bag, Sterile, Case Quantity 200
SDP
Catalog No. 5000670057
Encompass_Preferred
Audible "snap!" gives you the confidence that the tube is completely closed
Easily open or seal the tube with one hand; same hinged cap style as micro centrifuge tubes
Elongated lip on attached cap prevents finger contact with tube, reducing cross contamination
Ideal for sample storage, collection and experimental procedures
Single-piece design is easy to use in the lab or the field
Offers savings advantage versus traditional flip-top screw cap
Available in conical and self-standing bottoms
Catalog No. 50-006-70057 Supplier CELLTREAT Scientific Products Supplier No. 230425
Audible "snap!" gives you the confidence that the tube is completely closed
Easily open or seal the tube with one hand; same hinged cap style as micro centrifuge tubes
Elongated lip on attached cap prevents finger contact with tube, reducing cross contamination
Ideal for sample storage, collection and experimental procedures
Single-piece design is easy to use in the lab or the field
Offers savings advantage versus traditional flip-top screw cap
Available in conical and self-standing bottoms