Learn More
Chemglass Life Sciences 25mL Erlenmeyer Flask, 14/20 Outer Joint, Plain Ungraduated
SDP
Catalog No. CG154242
Encompass_Preferred
25mL Erlenmeyer Flask, 14/20 Outer Joint, Plain Ungraduated. Erlenmeyer flask with a standard taper outer joint. For stoppers see CG-3000.
Catalog No. CG154242 Supplier Chemglass Life Sciences Supplier No. CG154242
25mL Erlenmeyer Flask, 14/20 Outer Joint, Plain Ungraduated. Erlenmeyer flask with a standard taper outer joint. For stoppers see CG-3000.