Learn More
Chemglass Life Sciences 25mL Flowmeter, Bubble
SDP
Catalog No. CG160702
Encompass_Preferred
25mL Flowmeter, Bubble. Used for the control of carrier gases. Projection at top breaks bubbles. Supplied complete with rubber bulb. Hose connection has a O.D. of 9mm at the middle ring.
Catalog No. CG160702 Supplier Chemglass Life Sciences Supplier No. CG160702
25mL Flowmeter, Bubble. Used for the control of carrier gases. Projection at top breaks bubbles. Supplied complete with rubber bulb. Hose connection has a O.D. of 9mm at the middle ring.