Learn More
Chemglass Life Sciences Cylinder, Graduated, 25mL, 14/20 Outer Joint, "To Deliver"
SDP
Catalog No. CG122305
Encompass_Preferred
Cylinder, Graduated, 25mL, 14/20 Outer Joint. Cylinders are calibrated to contain at 20¿ C. Supplied without stopper. For standard taper stoppers see CG-3000. Tolerances and subdivisions are the same as CG-8242.
Catalog No. CG122305 Supplier Chemglass Life Sciences Supplier No. CG122305
Cylinder, Graduated, 25mL, 14/20 Outer Joint. Cylinders are calibrated to contain at 20¿ C. Supplied without stopper. For standard taper stoppers see CG-3000. Tolerances and subdivisions are the same as CG-8242.