Learn More
Research Products International Corp 25ml High Capacity Centrifuge Tubes, 8 bgs of 25, 200/pk
SDP
Catalog No. 502252995
Encompass
New 25mL tubes are ideal for any application that requires more than 15mL and less than 50mL, including midipreps, general sample prep, centrifugation, mixing and storage. Low profile, wide diameter format tubes allow faster re-suspension of pellets and 100% recovery of small samples. 25ml polypropylene tubes withstand up to 17,000xg.
Printed graduatioms, conical format
Withstand -86°C to 121°C
Sterile, RNase, DNase and Pyrogen free
Screw cap
This item is not returnable. View return policy
Catalog No. 50-225-2995 Supplier Research Products International Corp Supplier No. 248551
New 25mL tubes are ideal for any application that requires more than 15mL and less than 50mL, including midipreps, general sample prep, centrifugation, mixing and storage. Low profile, wide diameter format tubes allow faster re-suspension of pellets and 100% recovery of small samples. 25ml polypropylene tubes withstand up to 17,000xg.
Printed graduatioms, conical format
Withstand -86°C to 121°C
Sterile, RNase, DNase and Pyrogen free
Screw cap