Learn More
Chemglass Life Sciences 25mL Single Neck Round Bottom Flask (RBF) , Long Neck, 19/22 Outer Joint
SDP
Catalog No. CG151033
Encompass_Preferred
25mL Single Neck Round Bottomed Flask, Long Neck, 19/22 Outer Joint. Round bottom, HEAVY WALL single long neck flask with a standard taper outer joint.
Catalog No. CG151033 Supplier Chemglass Life Sciences Supplier No. CG151033
25mL Single Neck Round Bottomed Flask, Long Neck, 19/22 Outer Joint. Round bottom, HEAVY WALL single long neck flask with a standard taper outer joint.