Learn More
GREER LABORATORIES INC Mite House Dust Allergenic Extract, Sterile Solution, 25 mL
Catalog No. NC1487877
Encompass
GREER MITE HOUSE DUST Allergenic Extract is designed for the diagnosis and treatment of mite-induced allergic conditions. Formulated as a sterile solution with glycerin, it is suitable for percutaneous, intradermal, or subcutaneous administration. This standardized extract features a concentration range of 5,000 to 30,000 Allergy Units/mL, differentiating it with its clinically validated formulations sourced from Mite House Dust. Effective for managing allergy symptoms, such as asthma and rhinitis.
Catalog No. NC1487877 Supplier GREER LABORATORIES INC Supplier No. XPB81D3A25ML
GREER MITE HOUSE DUST Allergenic Extract is designed for the diagnosis and treatment of mite-induced allergic conditions. Formulated as a sterile solution with glycerin, it is suitable for percutaneous, intradermal, or subcutaneous administration. This standardized extract features a concentration range of 5,000 to 30,000 Allergy Units/mL, differentiating it with its clinically validated formulations sourced from Mite House Dust. Effective for managing allergy symptoms, such as asthma and rhinitis.