Learn More
Bellco Glass, Inc. 25mL Serological Pipet, Individual Paper/Plastic Wrapper Packed in Bags, Sterile
SDP
Catalog No. 502330229
Encompass_Preferred
All serological pipets are manufactured to the ASTM Standard Specification for Serological Pipets E-934Bi-directional (positive and negative) graduations are printed in bold easy-to-read inkPremium material barrier plugColor coded stripes and packaging enable quick volume identification Pack Description50 Individually Wrapped/BagCase Quantity200SterileYesNon-pyrogenicYesPrimary ColorClearSecondary ColorRedPrimary MaterialPolystyreneMaximum Volume25mLGraduatedYesGraduation Increments (Subdivisions)2/10mLRNase/DNase FreeYes
Catalog No. 50-233-0229 Supplier Bellco Glass, Inc. Supplier No. 229225B00
All serological pipets are manufactured to the ASTM Standard Specification for Serological Pipets E-934Bi-directional (positive and negative) graduations are printed in bold easy-to-read inkPremium material barrier plugColor coded stripes and packaging enable quick volume identification Pack Description50 Individually Wrapped/BagCase Quantity200SterileYesNon-pyrogenicYesPrimary ColorClearSecondary ColorRedPrimary MaterialPolystyreneMaximum Volume25mLGraduatedYesGraduation Increments (Subdivisions)2/10mLRNase/DNase FreeYes