Learn More
Bellco Glass, Inc. 25mL Pipet Individually
SDP
Catalog No. 502330389
Encompass_Preferred
All serological pipets are manufactured to the ASTM Standard Specification for Serological Pipets.Specifications Bidirectional positive and negative graduations are printed in bold black ink 10mL wide tip Serological pipet for viscous solutions. Wide tip is 4x larger than standard 10mL pipet 25 amp 50mL pipet features an antidrip tip for accurate delivery Color Coded stripes and packing enable quick volume identification Printed indicator arrow for quickly locating negative graduations when using a pipet controller. 10mL graduations 50 individually wrapped bag pack Case Quantity 200
Catalog No. 50-233-0389 Supplier Bellco Glass, Inc. Supplier No. 229025B00
All serological pipets are manufactured to the ASTM Standard Specification for Serological Pipets.Specifications Bidirectional positive and negative graduations are printed in bold black ink 10mL wide tip Serological pipet for viscous solutions. Wide tip is 4x larger than standard 10mL pipet 25 amp 50mL pipet features an antidrip tip for accurate delivery Color Coded stripes and packing enable quick volume identification Printed indicator arrow for quickly locating negative graduations when using a pipet controller. 10mL graduations 50 individually wrapped bag pack Case Quantity 200