Learn More
Bellco Glass, Inc. 25mL Short Serological Pipet, Individual Paper/Plastic Wrapper Packed in Bags, Sterile
SDP
Catalog No. 502329463
Encompass_Preferred
Compact design of short serological pipet provides ergonomic benefit and reduces storage spaceShorter length helps prevent touching pipet tip to surfaces in the hoodIdeal for students and those new to pipettingAll serological pipets are manufactured to the ASTM Standard Specification for Serological Pipets E-934Bi-directional (positive and negative) graduations are printed in bold easy-to-read inkPremium material barrier plugColor coded stripes and packaging enable quick volume identification Pack Description25 Individually Wrapped/BagCase Quantity100SterileYesNon-pyrogenicYesPrimary ColorClearSecondary ColorRedPrimary MaterialPolystyreneMaximum Volume25mLGraduatedYesGraduation Increments (Subdivisions)5/10mLRNase/DNase FreeYes
Catalog No. 50-232-9463 Supplier Bellco Glass, Inc. Supplier No. 229226B00
Compact design of short serological pipet provides ergonomic benefit and reduces storage spaceShorter length helps prevent touching pipet tip to surfaces in the hoodIdeal for students and those new to pipettingAll serological pipets are manufactured to the ASTM Standard Specification for Serological Pipets E-934Bi-directional (positive and negative) graduations are printed in bold easy-to-read inkPremium material barrier plugColor coded stripes and packaging enable quick volume identification Pack Description25 Individually Wrapped/BagCase Quantity100SterileYesNon-pyrogenicYesPrimary ColorClearSecondary ColorRedPrimary MaterialPolystyreneMaximum Volume25mLGraduatedYesGraduation Increments (Subdivisions)5/10mLRNase/DNase FreeYes