Learn More
Andwin Scientific 25ML PIPETTE REAG WELL 100/CS
SDP
Catalog No. NC0480183
Encompass_Preferred
The disposable pipetting reservoirs are made for multichannel pipetting applications. This product is compatible with any type of pipettors that are up to 12-channel width. This polystyrene reservoir has a unique trough construction, graduation marks on interior walls, and pour-off spouts on each corner. It has a volume capacity of 25 mL and has a dimension of 14.6 x 5.1 x 2.5 cm. This product is sold in a case of 100 white sterile reservoirs.
Catalog No. NC0480183 Supplier Andwin Scientific Supplier No. 89094664
The disposable pipetting reservoirs are made for multichannel pipetting applications. This product is compatible with any type of pipettors that are up to 12-channel width. This polystyrene reservoir has a unique trough construction, graduation marks on interior walls, and pour-off spouts on each corner. It has a volume capacity of 25 mL and has a dimension of 14.6 x 5.1 x 2.5 cm. This product is sold in a case of 100 white sterile reservoirs.