Learn More
Andwin Scientific 25ML REAGENT RESERVOIR 100/CS
SDP
Supplier:  Andwin Scientific 7049730
Encompass_Preferred
The 25 mL Reagent Reservoirs features a unique design of "Trough within a trough" that maximizes reagent recovery. It also has a wide base that gives stability and minimization of spills. There are graduated marks on inside wall that permit easy sample measurement. Pour-off spouts on all four corners are also provided to allow easy pouring out of samples. The reservoirs can be used with eight channel pipets on one side and one to four channel pipets on the other. This product is sold in a case of 100.
Catalog No. 50-930-354