Learn More
Bellco Glass, Inc. 25mL Reagent Reservoir, Bulk, Non-sterile
SDP
Catalog No. 502329320
Encompass_Preferred
reservoirs are uniquely treated to provide the lowest residual volume in the industry, meaning maximal reagent recovery, up to 67% better than comparable reservoirs.Trough-within-a-trough design to recover the “last few drops”Pour-off spouts at every cornerWider base for stabilityFree of animal productsSterile reservoirs are certified RNase/DNase free and non-pyrogenicWell-marked graduated increments on the inside wallBulk or individually wrapped packaging•Molded corner structures allow bulk packaged units to be easily separated for use•Clearly marked for recycling•Available in variety of sizes•12-channel polypropylene reservoir with lid is autoclavable
Catalog No. 50-232-9320 Supplier Bellco Glass, Inc. Supplier No. 451900214
reservoirs are uniquely treated to provide the lowest residual volume in the industry, meaning maximal reagent recovery, up to 67% better than comparable reservoirs.Trough-within-a-trough design to recover the “last few drops”Pour-off spouts at every cornerWider base for stabilityFree of animal productsSterile reservoirs are certified RNase/DNase free and non-pyrogenicWell-marked graduated increments on the inside wallBulk or individually wrapped packaging•Molded corner structures allow bulk packaged units to be easily separated for use•Clearly marked for recycling•Available in variety of sizes•12-channel polypropylene reservoir with lid is autoclavable