Learn More
Andwin Scientific Sterile Reagent Reservoir with Divider, 25mL, 100/cs
SDP
Catalog No. NC1659398
Encompass
Sterile Reagent Reservoir with Divider, 25mL, 100/cs, divider allows for pipetting with eight channels on one side and four channels or a single channel on the other
Catalog No. NC1659398 Supplier Andwin Scientific Supplier No. RESE2000
Sterile Reagent Reservoir with Divider, 25mL, 100/cs, divider allows for pipetting with eight channels on one side and four channels or a single channel on the other