Learn More
MTC Bio Lids for 25mL solution reservoirs, sterile, 5 per bag
Catalog No. NC2017619
Encompass_Preferred
These new reservoir lids provide an added measure of security and protection for valuable reagents. Made of polystyrene, the lids are custom formed to create a perfect perimeter around MTC Bio’s 25mL reservoirs. This 360 degree protection inhibits evaporation, while also protecting solutions/reagents from dust and contamination. Lids can be purchased in sterile packs of 5 and are in-stock now. Subject to change without notice.
Catalog No. NC2017619 Supplier MTC Bio Supplier No. P8025LID
These new reservoir lids provide an added measure of security and protection for valuable reagents. Made of polystyrene, the lids are custom formed to create a perfect perimeter around MTC Bio’s 25mL reservoirs. This 360 degree protection inhibits evaporation, while also protecting solutions/reagents from dust and contamination. Lids can be purchased in sterile packs of 5 and are in-stock now. Subject to change without notice.